Learn More
Microbiologics™ EZ-CFU™ Aspergillus brasiliensis ATCC™ 16404™
Description
Includes
Product warranty and peel-off certificate of assay
Lyophilized microorganism preparations delivering the desired 10 to 100CFU/0.1mL of challenge material (following hydration and dilution of 1.0mL of hydrated suspension in 9.0mL of pH 7.2 phosphate buffer).
- Traceable to reference cultures
Order Info
Supplied as two vials of a single assayed microorganism (10 lyophilized pellets in each vial) and 10 vials of hydrating fluid (2.0mL in each vial)
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
Specifications
Specifications
| ATCC® | 16404 |
| Format | EZ-CFU |
| Product Type | EZ-CFU |
| For Use With (Application) | Assayed microorganism preparations for USP quantitative microorganism challenges. |
| Includes | Product warranty and peel-off certificate of assay |
| Packaging Type | Vial |
| Species | Aspergillus brasiliensis |