Learn More
Microbiologics™ EZ-CFU™ Bacillus subtilis ATCC™ 6633™
Product Code. 11901426
Encompass_Preferred
Change view
Click to view available options
Species:
B. subtilis subsp. spizizenni
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Species Unit Size
11901426 B. subtilis subsp. spizizenni Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11901426 Supplier Microbiologics™ Supplier No. 0486C
 Restricted Item
Spedizione stimata: 07-12-2026
Includes
Product warranty and peel-off certificate of assay
Assayed microorganism preparation for USP quantitative microorganism challenges
Lyophilized microorganism preparations delivering the desired 10 to 100CFU/0.1mL of challenge material (following hydration and dilution of 1.0mL of hydrated suspension in 9.0mL of pH 7.2 phosphate buffer).
  • Traceable to reference cultures
Order Info
Supplied as two vials of a single assayed microorganism (10 lyophilized pellets in each vial) and 10 vials of hydrating fluid (2.0mL in each vial)
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
Specifications
ATCC® 6633
Format EZ-CFU
Product Type EZ-CFU
For Use With (Application) Assayed microorganism preparations for USP quantitative microorganism challenges.
Includes Product warranty and peel-off certificate of assay
Packaging Type Vial
Species B. subtilis subsp. spizizenni