Learn More
Microbiologics™ EZ-CFU™ Escherichia coli ATCC™ 8739™
Codice prodotto. 11901406
Encompass_Preferred
Cambia vista
Click to view available options
Specie:
Escherichia coli
Dimensione della confezione:
pezzi 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Specie Dimensione della confezione
11901406 Escherichia coli pezzi 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11901406 Fornitore Microbiologics™ N. del fornitore 0483C
 Articolo soggetto a restrizioni
Spedizione stimata: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
Includes
Product warranty and peel-off certificate of assay
Assayed microorganism preparation for USP quantitative microorganism challenges
Lyophilized microorganism preparations delivering the desired 10 to 100CFU/0.1mL of challenge material (following hydration and dilution of 1.0mL of hydrated suspension in 9.0mL of pH 7.2 phosphate buffer).
  • Traceable to reference cultures
Order Info
Supplied as two vials of a single assayed microorganism (10 lyophilized pellets in each vial) and 10 vials of hydrating fluid (2.0mL in each vial)
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
Specifications
ATCC® 8739
Formato EZ-CFU
Product Type EZ-CFU
Da utilizzare con (applicazione) Assayed microorganism preparations for USP quantitative microorganism challenges.
Include Product warranty and peel-off certificate of assay
Tipo di imballaggio Vial
Specie Escherichia coli
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.