Learn More
Staphylococcus epidermidis ATCC™ 12228™†, Microbiologics™
Descrizione
KWIK-STIK™
- Clever design and accurate, repeatable results
- Each device features a single strain of Staphylococcus epidermidis in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
- For added convenience, a peel-off identification label is included for easy documentation
- Available in packs of 2, 6 or 1
- The most economical reference stock culture option; lyophilized pellet containing a single strain of Staphylococcus epidermidis
- Easy to use; simply re-hydrate the pellet and inoculate
- Available in a re-sealable vial containing 6 or 10 pellets
- ATCC™ 12228™†
Specifica
Specifica
| ATCC® | ATCC 12228 |
| Livello di biosicurezza | 1 |
| Certificazione/Conformità | CE, online certificate of analysis |
| Content And Storage | 2°C to 8°C |
| Forma | Lyophilized Pellet |
| Formato | KWIK-STIK |
| Tipo di media | Kwik-stik™ |
| Product Type | Quality Control Organism |
| Da utilizzare con (applicazione) | Culture purpose and QC application |
| Quantità | 2/Pk. |
| Vedi altri risultati |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.