Learn More
Vileda Professional™ MicroControl Controlled Environments Mop
Description
- 100% microfiber mop surface offering high cleaning performance
- Patented edge cleaning lip design allows for effective cleaning of corners, skirt boards etc.
- Straight and clean design with no dirt traps
- Multilayer design with sealed edges for high absorbency, low particle shedding and top stability
- Ergonomic performance and high efficiency due to the Vileda™ pre-prepared method
- Swap mopping system: One cleaning tool for floors, walls and ceilings
- Ideal microfiber cleaning results have been independently tested and validated
Specifications
Specifications
| Product Type | Mop Pad |
| Color Code | Blue |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Color-coded | No |
| Material | Polyester |
Product Title
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?