Learn More
Vileda Professional™ MicroEffective Controlled Environments Mop
Codice prodotto. 17842496
Cambia vista
Click to view available options
Materiale:
Polyester
Dimensione della confezione:
pezzi 30
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Materiale Dimensione della confezione
17842496 Polyester pezzi 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 17842496 Fornitore Vileda Professional™ N. del fornitore 161533
  • ONLINE EXCLUSIVE PRICE
    1175.00€
    1017.35€ / pezzi 30
    Risparmia 157.65€
    13% Sconto
Spedizione stimata: 21-12-2026
MicroEffective CE Mop is a reusable concept with strong cleaning performance and low friction to clean less strict classification rooms. Compatible in ISO 8 to 9 areas with an improved version applicable for wider ISO applications (ISO 7 to 9).
  • Efficient polyethersulfone-microfiber mop-cleaning performance coupled with extended coverage
  • Hygienic, externally proven bacterial removal on floor, wall and ceiling surfaces
  • Very low particle shedding of microfiber mops (Cat 1 for Hoods per IEST-RP-CC03.4); up to ISO 5 (GMP A/B)
  • Enhanced decontamination result based on open side corners in the pockets
  • Approved for CE usage/Designed for Sterilization (autoclaving/gamma irradiation)
  • Ergonomic easy gliding mops and light weight mopping system
Specifica
Product Type Mop Pad
Codice colore Gray
Sterilità Non-sterile
Autoclavabile Sterilizzabile in autoclave
Monouso Reusable
Color-coded No
Materiale Polyester