Learn More
Epredia™ Microm™ Cryostats Heat Extractor
Product Code. 12033059 Shop All Epredia Products
Change view
Click to view available options
For Use With (Equipment):
HM 525
HM 550 and 560
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12033059 HM 550 and 560 Each
12023059 HM 525 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12033059 Supplier Epredia™ Supplier No. 524510
  • ONLINE EXCLUSIVE PRICE
    145.00€
    127.98€ / pezzi 1
    Risparmia 17.02€
    11% Sconto
Spedizione stimata: 30-11-2026
Use Epredia™ Microm™ Cryostats Heat Extractors with Epredia™ Cryostats.
  • Multipositional heat extractor is incorporated on a vertical shaft for even freezing and easy removal from specimen
  • Can be operated from any position on the freeze bar
  • Freezing time is shortened if freezing stage and tissue are placed on an active Peltier
TRUSTED_SUSTAINABILITY
Specifications
Description Heat Extractor for HM560
Product Type Heat Extractor
For Use With (Equipment) HM 550 and 560