Learn More
Simport™ Scientific Micromesh™ Biopsy Processing/Embedding Cassette in Quickload™ stacks for Primera printers
Secure specimens during processing with Simport Scientific™ Micromesh™ Biopsy Processing/Embedding Cassette. No need for biopsy pads thanks to the 1676 square 0.36mm openings of the mesh.
Marca: Simport™ Scientific M407-6T
Dettagli aggiuntivi : Peso : 5.30900kg
Descrizione
In Quickload™ stacks, for Primera printers
- 45° Anterior writing area
- 1676 square opening (0.36 mm)
- No biopsy pads needed
- 1 large 27 mm compartment
- Recessed cover
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifica
2000 | |
Blue | |
Acetal |