Learn More
Motic™ Microscope Head, SMZ-171-BH
Codice prodotto. 15485502
Cambia vista
Click to view available options
Potenza di ingrandimento:
0.75X to 5X
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Potenza di ingrandimento Dimensione della confezione
15485502 0.75X to 5X Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 15485502 Fornitore Motic™ N. del fornitore 1101000902961
  • ONLINE EXCLUSIVE PRICE
    842.00€
    753.39€ / pezzi 1
    Risparmia 88.61€
    10% Sconto
Spedizione stimata: 14-01-2027
Optically superior stereo zoom microscope head for the SMZ-171 microscope. Motic™ Microscope Head, SMZ-171-BH is a replacement head for the versatile and powerful SMZ-171 microscope.
  • Head only.
  • 60° binocular stereo head.
  • Wide field adjustable eyepieces 10S/F.N. 23.
  • 6.7:1 zoom ratio and WD = 110mm.
  • Magnification range: 0.75x - 5x.
  • Electrostatic discharge.
Specifica
Include Electrostatic discharge feature
Potenza di ingrandimento 0.75X to 5X
Product Type Binocular stereomicroscope head
Eyepiece WF 10X/F.N. 23, Adjustable