Learn More
Applied Biosystems™ MicroSEQ™ ID 16S rDNA 500 Library v2022
Sfoglia Tutto Applied Biosystems ProdottiDescrizione
The MicroSEQ ID 16S rDNA 500 Library v2022 is for use with MicroSEQ ID Microbial Identification Software v4.0. It contains over 2100 reference 16S rRNA sequences from bacteria. The library entries contain the first 500 bases of the 16S rRNA gene, enough to identify bacterial unknowns.
Specifica
Specifica
| Sequencing Type | Genome and DNA Sequencing |
| Da utilizzare con (apparecchiatura) | SeqStudio Genetic Analyzers, SeqStudio Flex Genetic Analyzers |
| Da utilizzare con (applicazione) | Microbial Identification |
| Licenza | Static |
| Sistema operativo | Microsoft Windows 10 |
| Linea di prodotti | MicroSeq™ ID |
| Product Type | Microbial Identification Software |
| Quantità | 1 Ea. |
| Software | Data Collection and Analysis Software |