Learn More
Applied Biosystems™ MicroSEQ™ ID Fungal Gene Library v2022
Sfoglia Tutto Applied Biosystems ProdottiDescrizione
The MicroSEQ ID Fungal Gene Library v2022 is for use with MicroSEQ ID Microbial Identification Software v4.0. It contains over 1300 reference sequences from the D2 region of the Large Subunit (rDNA) of yeast and mold species against which an unknown yeast or mold sequence can be compared in order to identify the unknown organism.
Specifica
Specifica
| Sequencing Type | Genome and DNA Sequencing |
| Da utilizzare con (apparecchiatura) | SeqStudio Genetic Analyzers, SeqStudio Flex Genetic Analyzers |
| Da utilizzare con (applicazione) | Microbial Identification |
| Licenza | Static |
| Sistema operativo | Microsoft Windows 10 |
| Linea di prodotti | MicroSeq™ ID |
| Product Type | Microbial Identification Software |
| Quantità | 1 Ea. |
| Software | Data Collection and Analysis Software |