Learn More
Labconco™ Microwell Plate Holder
Product Code. 15868684 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
FreeZone™ Freeze Dryers
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15868684 FreeZone™ Freeze Dryers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15868684 Supplier Labconco™ Supplier No. 7369600
  • ONLINE EXCLUSIVE PRICE
    280.00€
    227.30€ / pezzi 1
    Risparmia 52.70€
    18% Sconto
 Restricted Item
Spedizione stimata: 09-02-2027
Solid aluminum base tightly holds microplates.
Microwell Plate Holder
  • Made of solid aluminum
  • Eliminates air gap on bottom of plate for excellent heat transfer to microplate
  • Gives temperature stability to small samples that are susceptible to meltback
  • Microplate required and not included
Specifications
Type Microplate Holder
Material Aluminum
For Use With (Equipment) FreeZone™ Freeze Dryers
Includes Solid aluminum block that holds microwell plate. Microwell plate not included.
Width (English) 5.7 in.
Height (English) 0.7 in.
Height (Metric) 1.78 cm
Length (English) 4 in.
Length (Metric) 10.16 cm
Shipping Weight (English) 2 lb.
Weight (English) 1.3 lb.