Learn More
Miglitol, MedChemExpress
Product Code. 16480948
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480948 5 g 5g
16412199 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16480948 Supplier Medchem Express Supplier No. HYB0481
 Restricted Item
Spedizione stimata: 27-11-2026
MedChemExpress Miglitol is an oral anti-diabetic drug that acts by inhibiting the ability of the patient to breakdown complex carbohydrates into glucose.
Miglitol is an oral anti-diabetic drug that acts by inhibiting the ability of the patient to breakdown complex carbohydrates into glucose.
Specifications
Chemical Name or Material Miglitol
Physical Form Solid
CAS 72432-03-2
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C8H17NO5
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym BAY1099 BAY-m1099
Solubility Information H2O : ≥ 200 mg/mL (965.16 mM)
SMILES O[C@@H]1[C@@H](CO)N(CCO)C[C@H](O)[C@H]1O
Molecular Weight (g/mol) 207.22
Formula Weight 207.22
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only