Learn More
Thermo Scientific™ Mini-phone Jack/USB cable for Eutech™ 450 series/2700 series meters
Product Code. 15397073
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
15397073 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15397073 Supplier Thermo Scientific™ Supplier No. MINIPHONEUSB
  • ONLINE EXCLUSIVE PRICE
    39.15€
    31.50€ / pezzi 1
    Risparmia 7.65€
    19% Sconto
Spedizione stimata: 01-12-2026
Eutech™ provides various types of cables and adapters available for use with Eutech™ meters.
Eutech™ provides various types of cables and adapters available for use with Eutech™ meters. The Mini-phone Jack, 2.5mm /USB cable is used for Eutech 450/ 2700 series meters.
Features:
  • USB cable, 2.5mm mini phone jack to USB cable
  • Used with: Eutech 450 / 2700 meters
TRUSTED_SUSTAINABILITY
Specifications
Description USB cable, mini phone plug to USB
Type Mini-Phone Jack/USB Cable