Descrizione
Prevent backflow to protect your equipment. Work with both liquids and gases. Use for pressure or vacuum applications. PVDF and Viton construction provides excellent corrosion and chemical resistance. 0.18 psi breaking pressure
Specifica
Specifica
| Dimensioni dei tubi | 0.3125 in. |
| Capacità del flusso | 0.12 |
| Ingresso | 3/8 in. hose barb |
| Porta di uscita | Barbed |
| Materiale (guarnizione) | Viton |