Learn More
mitochondrial ribosomal protein L30 Antibody, Novus Biologicals™
Codice prodotto. 18609975 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
100 μg
50 μg
Dimensione della confezione:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18609975 100 μg 100µL
18648485 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 18609975 Fornitore Novus Biologicals N. del fornitore NBP305921100ul
Rabbit Polyclonal Antibody
mitochondrial ribosomal protein L30 Polyclonal antibody specifically detects mitochondrial ribosomal protein L30 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifica
Antigene mitochondrial ribosomal protein L30
Applicazioni Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classificazione Polyclonal
Coniugato Unconjugated
Diluizione Western Blot 1:500-1:2000, Immunoprecipitation 1-5uL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200
Formulazione PBS, pH7.0
Alias gene 39S ribosomal protein L30, mitochondrial, L28mt, L30mt, mitochondrial ribosomal protein L30, MRP-L28FLJ44438, MRPL28MGC3314,39S ribosomal protein L28, mitochondrial, MRP-L30, RPML28MGC24095
Specie ospite Rabbit
Immunogeno Produced in rabbits immunized with E. coli-derived Human mitochondrial ribosomal protein L30 fragment.
Metodo di purificazione Antigen and protein A Affinity-purified
Quantità 100 μg
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 51263
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Forma Purified
Isotype IgG