Learn More
Mitoxantrone dihydrochloride, MedChemExpress
Product Code. 16461877
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461877 50 mg 50mg
16456206 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16461877 Supplier Medchem Express Supplier No. HY13502A
 Restricted Item
Spedizione stimata: 30-11-2026
MedChemExpress Mitoxantrone dihydrochloride is a potent topoisomerase II inhibitor.
Mitoxantrone dihydrochloride also inhibits protein kinase C (PKC) activity with an IC50 of 8.5 μM. Mitoxantrone dihydrochloride induces apoptosis of B-CLL (B-chronic lymphocytic leukaemia) cells. Mitoxantrone dihydrochloride shows antitumor activity. Mitoxantrone dihydrochloride also has anti-orthopoxvirus activity with EC50s of 0.25 μM and and 0.8 μM for cowpox and monkeypox, respectively.
Specifications
Chemical Name or Material Mitoxantrone dihydrochloride
CAS 70476-82-3
Color Dark blue
Purity Grade Notes Research
Molecular Formula C22H30Cl2N4O6
Quantity 50 mg
Synonym Mitozantrone dihydrochlorideNSC 301739 dihydrochloride
Solubility Information H2O : 100 mg/mL (193.27 mM; Need ultrasonic) ∣DMSO : ≥ 43 mg/mL (83.11 mM)
SMILES O=C1C2=C(C(NCCNCCO)=CC=C2NCCNCCO)C(C3=C(O)C=CC(O)=C13)=O.[H]Cl.[H]Cl
Molecular Weight (g/mol) 517.4
Formula Weight 517.4
Percent Purity 99.55%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only