Learn More
Mixed Bed Exchange Resin, IONAC™ NM-60 H+/OH- Form, Type I, Strong acid/strong base; sulfonated/alkyl quaternary ammonium polystyrenes, Beads, 16 to 50 Mesh, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Marca: JT Baker 4631-01
328.70 EUR valido fino al 2024-12-31
Usa il codice promo "23268" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.99000kg
Specifica
16 to 50 Mesh | |
500 g | |
Mixed Bed Exchange Resin, IONAC™ NM-60 H+/OH- Form, Type I | |
Beads |
Wide Mouth Amber Glass Bottle | |
1L = 1.20kg | |
Strong acid/strong base; sulfonated/alkyl quaternary ammonium polystyrenes | |
BAKER ANALYZED™ Reagent |