Learn More
Fisherbrand™ Mixed Heating (Steam or Electrical)
Mixed heating from a steam source for water heating and an electrically powered source for drying.
Brand:  Fisherbrand™
Additional Details : Peso : 0.10000kg
 View more versions of this product
Product Code. 17117834
  • 2948.45€ / Pezzo
Spedizione stimata: 28-02-2023
per visualizzare lo stock disponibile
Add to basket
Description
Description
  • The energy for water heating comes from steam source
  • The energy for drying is electrically powered
  • If the steam fails, the machine continues to run on electrical power
  • The machine can be programmed to run on steam in certain periods and electricity at others
Specifications
Specifications
Steam or Electrical Heating
Blue Line 6 / IQ6
Videos
SDS
Documents
Documents
Product Certifications
Special Offers
Special Offers