Learn More
Fisherbrand™ Mixed Heating (Steam or Electrical)
Mixed heating from a steam source for water heating and an electrically powered source for drying.
Marca:  Fisherbrand™
Dettagli aggiuntivi : Peso : 0.30000kg
Codice prodotto. 17117794
  • 2566.95€ / Pezzo
Spedizione stimata: 28-02-2023
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • The energy for water heating comes from steam source
  • The energy for drying is electrically powered
  • If the steam fails, the machine continues to run on electrical power
  • The machine can be programmed to run on steam in certain periods and electricity at others
Specifica
Specifica
Steam or Electrical Heating
Blue Line 5 / IQ5