Descrizione
Treat unsoftened tap water or a TIS of more than 10ppm with the Thermo Scientific™ Mixed Multi Mini Water Softener. For use with Thermo Scientific™ Barnstead™ water purification systems, it is required if feed water silt density index (SDI) is greater than 3.
Required for use: Softener salt, hardness detection kit and 5μm filter with carbon. This softener is ideal for use with the Barnstead LabTower™ EDI water purification system.
Specifica
Specifica
| Descrizione | Mix Multi Mini Water softener |
| Da utilizzare con (apparecchiatura) | LabTower EDI |
| Tensione | 230 V |