missing translation for 'onlineSavingsMsg'
Learn More

MK-3207, MedChemExpress

Codice prodotto. p-200099418
Encompass_Preferred
Cambia vista
Click to view available options
Quantità:
5 mg
10 mg
Dimensione della confezione:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità unitSize
16483768 10 mg 10mg
16426497 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16483768 Supplier Medchem Express Supplier No. HY10301

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

MedChemExpress MK-3207 is a potent and orally bioavailable CGRP receptor antagonist (IC50= 0.12 nM; Ki= 0.024 nM); highly selective versus human AM1, AM2, CTR, and AMY3.

MK-3207 is a potent and orally bioavailable CGRP receptor antagonist (IC50= 0.12 nM

Specifications

Materiale o nome chimico MK-3207
CAS 957118-49-9
Colore White
Note sul grado di purezza Research
Formula molecolare C31H29F2N5O3
Quantità 10 mg
Informazioni di solubilità DMSO : ≥ 150 mg/mL (269.01 mM)
SMILES O=C1C2(NC[C@@H](C3=CC(F)=CC(F)=C3)N1CC(NC4=CC5=C(C[C@@]6(C(NC7=C6C=CC=N7)=O)C5)C=C4)=O)CCCC2
Molecular Weight (g/mol) 557.59
Peso formulazione 557.59
Percent Purity 96.47%
Grado Research
Forma fisica Solid
Da utilizzare con (applicazione) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.