missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VistaLab™ Technologies MLA™ Silver Brand Precision Pipetters
Made of precision engineered aluminum alloy and stainless steel with limited Lifetime Warranty (covering parts). Individually serialized with Calibration Certificate included.
Marca: VistaLab™ Technologies 1119
Dettagli aggiuntivi : Peso : 0.50000kg
Descrizione
- “To deliver” capability ensures volume accuracy over a wide range of viscosities
- Ideal for repetitive delivery of reagents when high precision and accuracy are required
- Capacity engraved on bonnet
Specifica
±1% | |
Silver | |
MLA Precision | |
Non-sterile | |
450 μL | |
±0.44% | |
Limited Lifetime | |
2 in. |
Not Autoclavable | |
Aluminum Alloy, Stainless Steel | |
1 | |
Precision Pipetter | |
Nozzle Insert, Calibration Key | |
1 Ea. | |
9 in. | |
1.5 in. |