missing translation for 'onlineSavingsMsg'
Learn More

MLANA Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 18472-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898494

  • 129.00€ / 20µL

Spedizione stimata: 14-12-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

MART1 is involved in melanosome biogenesis by ensuring the stability of GPR143. Plays a vital role in the expression, stability, trafficking, and processing of melanocyte protein PMEL, which is critical to the formation of stage II melanosomes.
Specifica

Specifica

MLANA
Polyclonal
Unconjugated
MLANA
Antigen LB39 AA, Antigen SK29 AA, MART 1, MART1, melan A, MLANA, Protein Melan A
Rabbit
Antigen Affinity Chromatography
RUO
2315, 77836
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q16655
Mlana
MLANA Fusion Protein Ag13346
20 μL
Primary
Mouse, Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali