Learn More
Macherey-Nagel™ MN 652 Grade Filter Papers, Round
Wet strengthened for use in industrial applications. Macherey-Nagel™ MN 652 Grade Filter Papers, Round are creped technical filter papers with fast filtration speed for use with essential oils, gelatine, resin solutions, sugared spirits, syrups, and transformer/turbine oil regeneration.
Marca: Macherey-Nagel™ 446018
Descrizione
High quality MN filter papers use cotton linters and refined pulp with a high level of α-cellulose. Cotton linters are short-fiber seed hairs from cotton seeds, which cannot be used for textile purposes, but which are highly suitable for the manufacture of soft and absorbent filter papers. In addition to cotton linters, Macherey-Nagel uses mainly pulp, which is obtained by chemical treatment of plant materials, e.g. coniferous or deciduous wood.
Specifica
| 140 g/m2 | |
| Round | |
| 15sec. | |
| MN 652 | |
| 0.45 mm | |
| 185 mm |
| Creped, Wet-strengthened | |
| Cellulose | |
| Technical, industrial applications (e.g. essential oils, gelatine, resin solutions, spirits, syrups, and transformer/turbine oil regeneration) | |
| 100 Pack | |
| Filter Paper |