missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MND1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11636-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Required for proper homologous chromosome pairing and efficient cross-over and intragenic recombination during meiosis. Stimulates both DMC1- and RAD51-mediated homologous strand assimilation, which is required for the resolution of meiotic double-strand breaks.Specifica
MND1 | |
Polyclonal | |
Unconjugated | |
MND1 | |
GAJ, MND1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
76915, 84057 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8K396, Q9BWT6 | |
Mnd1 | |
MND1 Fusion Protein Ag2210 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |