Learn More
Hamilton™ 1000 Series Gastight Instrument Syringes
With a precision-machined PTFE plunger tip which creates a leak-free seal.
Marca:  Hamilton™ 208643
Dettagli aggiuntivi : Peso : 0.20000kg
Codice prodotto. 15335104
  • 160.00€ / Pezzo
Spedizione stimata: 21-05-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • Tip wipes the interior of the syringe barrel free of sample
  • Useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze
  • Gas sterilizable
Specifica
Specifica
2.5 mL