Learn More
Ansell Model 192-G01 - AlphaTec™ 3000 Ultrasonically Welded and Taped Coverall
Shop All Ansell ProductsDescription
- Multi-layer barrier fabric provides reliable protection against a wide range of chemicals.
- Innovative safety features, including a dual-zip system, enhance protection.
- Lightweight yet durable for comfortable wear.
- Bright yellow high-visibility color helps improve worker safety.
- Anti-static and tested in accordance with EN 1149-5.
Specifications
Specifications
| Product Type | Hooded Coveralls |
| Size | Medium |
| Gender | Unisex |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Yellow |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Ties |
| Cuff Style | Locking Glove Ring |
| Neck Style | Hooded |
| Show More |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?