Learn More
Monoisodecyl Phthalate, TRC
Codice prodotto. 30132447
Cambia vista
Click to view available options
Quantità:
10 mg
25 mg
Dimensione della confezione:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30132447 25 mg 25mg
30003688 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 30132447 Fornitore Toronto Research Chemicals N. del fornitore TRCM54790525MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 69725-01-5
Formula molecolare C18 H26 O4
Molecular Weight (g/mol) 306.4
Sinonimo 1,2-Benzenedicarboxylic Acid Monoisodecyl Ester, Phthalic Acid Isodecyl Ester
IUPAC Name 2-(8-methylnonoxycarbonyl)benzoic acid
SMILES OC(C1=CC=CC=C1C(OCCCCCCCC(C)C)=O)=O
Specifica
Materiale o nome chimico Monoisodecyl Phthalate
Note sul grado di purezza HPLC
Quantità 25 mg
InChI Formula InChI=1S/C18H26O4/c1-14(2)10-6-4-3-5-9-13-22-18(21)16-12-8-7-11-15(16)17(19)20/h7-8,11-12,14H,3-6,9-10,13H2,1-2H3,(H,19,20)
Peso formulazione 306.1831
Percent Purity >95%