Learn More
Monopropyl Phthalate O-Beta-D-Glucuronide, TRC
Product Code. 30011500
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011500 5 mg 5mg
30133695 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30011500 Supplier Toronto Research Chemicals Supplier No. TRCM5671055MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
Molecular Formula C17H20O10
Molecular Weight (g/mol) 384.33
Synonym 1-(Propyl 1,2-Benzenedicarboxylate} β-D-Glucopyranuronic Acid;
IUPAC Name (2S,3S,6S)-3,4,5-trihydroxy-6-((2-(propoxycarbonyl)benzoyl)oxy)tetrahydro-2H-pyran-2-carboxylic acid
SMILES O=C(OCCC)C1=CC=CC=C1C(O[C@@H]2O[C@H](C(O)=O)[C@@H](O)C(O)C2O)=O
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C17H20O10/c1-2-7-25-15(23)8-5-3-4-6-9(8)16(24)27-17-12(20)10(18)11(19)13(26-17)14(21)22/h3-6,10-13,17-20H,2,7H2,1H3,(H,21,22)/t10?,11-,12?,13-,17-/m0/s1
Formula Weight 384.11
Chemical Name or Material Monopropyl Phthalate O-beta-D-Glucuronide