Learn More
Pro-Lab Diagnostics™ Monovalent Flagella (H) Salmonella Antisera
Product Code. 11969124
Change view
Click to view available options
Grade:
Antigen Factor 2
Antigen Factor 5
Antigen Factor d
Antigen Factor eh
Antigen Factor enx
Antigen Factor f
Antigen Factor gm
Antigen Factor h
Antigen Factor k
Antigen Factor l, v
Antigen Factor l, w
Antigen Factor q
Antigen Factor s
Antigen Factor t
Antigen Factor u
Antigen Factor z13
Antigen Factor z24
Antigen Factor z29
Antigen Factor z32
Antigen Factor z6
Unit Size:
Each
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Grade Unit Size
11969124 Antigen Factor z6 Each
11999084 Antigen Factor eh Each
11979084 Antigen Factor d Each
11929094 Antigen Factor h Each
11959094 Antigen Factor gm Each
11989094 Antigen Factor u Each
11999094 Antigen Factor s Each
11919104 Antigen Factor t Each
11929104 Antigen Factor f Each
11939104 Antigen Factor q Each
11959104 Antigen Factor k Each
11979104 Antigen Factor l, w Each
11989104 Antigen Factor l, v Each
11919114 Antigen Factor z13 Each
11989114 Antigen Factor z24 Each
11999114 Antigen Factor z32 Each
11919124 Antigen Factor z29 Each
11929124 Antigen Factor 2 Each
11939124 Antigen Factor 5 Each
11909094 Antigen Factor enx Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11969124 Leverantör Pro-Lab Diagnostics™ Leverantörsnummer PL.6157
 Restricted Item
In stock
Lägg i varukorgen
Edge
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy
Use in slide agglutination testing for serotyping of Salmonella isolates according to Kauffmann-White classification. Pro-Lab Diagnostics™ Monovalent Flagella (H) Salmonella Antisera is a comprehensive range of monovalent Salmonella antigens delivered in economical 3.0mL bottles.
While polyvalent O (somatic) antisera aid in initial serogrouping, full identification of O antigens is achieved using monovalent-specific O antisera. The serotype of Salmonella isolates can then be determined using H (flagella) antisera.
  • Ready-to-use diluted antisera with 0.01% thimerosal as a preservative
  • Produced using reference strains prepared by World Health Organization guidelines
  • Certified reference center performance
  • Presented in color-coded dropper bottles
  • Long shelf life: up to 5 years from date of manufacture
Specifikationer
Certifications/Compliance W.H.O.
Content And Storage 2°C to 8°C, Do Not Freeze
Grade Antigen Factor z6
Product Type Antiserum
For Use With (Application) Traditional Slide Agglutination Testing
Packaging Type Dropper Bottle
Quantity 3.0 mL
Slide Type Agglutination
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.