missing translation for 'onlineSavingsMsg'
Learn More

MOPS, 0.5M buffer soln., pH 9.0, Thermo Scientific™

Catalog Number p-7041981
Click to view available options
Quantità:
100 mL
250 mL
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1132-61-2
C7H15NO4S
209.26
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
70807
CHEBI:44115
3-morpholin-4-ylpropane-1-sulfonic acid
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1132-61-2
C7H15NO4S
209.26
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
70807
CHEBI:44115
3-morpholin-4-ylpropane-1-sulfonic acid
[O-]S(=O)(=O)CCC[NH+]1CCOCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico MOPS buffer soln.
Concentrazione 0.5 M
pH 9.0
Forma fisica Liquid
Quantità 100 mL
Odore Odorless
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'