missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse anti-Bovine IgG2 Secondary Antibody, HRP
Descrizione
Mouse anti Bovine IgG2, clone IL-A73, is a monoclonal antibody specific for bovine IgG2 and does not recognise other bovine immunoglobulin classes.
Anti-Bovine secondary antibodies are affinity-purified antibodies with well-characterized specificity for bovine (cow, ox) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifica
Specifica
| Antigene | Bovine IgG2 |
| Applicazioni | ELISA |
| Classificazione | Monoclonal |
| Clone | IL-A73 |
| Concentrazione | 1 mg/mL |
| Coniugato | HRP |
| Formulazione | PBS with 0.01% thimerosal |
| Specie ospite | Mouse |
| Immunogeno | Purified bovine IgG2. |
| Metodo di purificazione | Protein G |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?