Learn More
CD127 Mouse anti-Human, APC-R700, Clone: HIL-7R-M21, BD
Descrizione
The hIL-7R-M21 monoclonal antibody specifically binds to the 60-90 kDa glycoprotein, CD127. CD127 is also known as the IL-7 receptor alpha (IL-7R α ) subunit. The IL-7 receptor complex is a heterodimer composed of CD127 and the common gamma chain ( γc, CD132), shared by other cytokine receptors (IL-2R, IL-4R, IL-9R, IL-15R, and IL-21R). CD127 is expressed on thymocytes, T- and B-cell progenitors, mature T cells, and some lymphoid and myeloid cells. In vitro experiments show the expression of CD127 is down-regulated following T cell activation. Studies indicate that the IL-7 Receptor plays an important role in the proliferation and differentiation of mature T cells. Recently, it has been shown that low surface expression of CD127, in combination with intermediate to high surface expression of CD25, the α chain of the IL-2 receptor complex, can distinguish between human regulatory and conventional CD4+ T cells in human adult and cord blood, lymph nodes and thymus.
Specifica
Specifica
| Antigene | CD127 |
| Applicazioni | Flow Cytometry |
| Classificazione | Monoclonal |
| Clone | HIL-7R-M21 |
| Coniugato | APC-R700 |
| Descrizione | IL-7R; IL7R; IL7RA; IL-7Rα; IL-7R-alpha; Interleukin-7 Receptor alpha |
| Formulazione | Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide. |
| Specie ospite | Mouse |
| Immunogeno | Recombinant Human IL-7R |
| Metodo di purificazione | Affinity Purified |
| Vedi altri risultati |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.