missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse anti-Human Lambda Light Chain Secondary Antibody
Descrizione
This antibody is not recommended for use in paraffin embedded tissue sections A suggested positive control for this product is human B cells.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifica
Specifica
| Antigene | Human Lambda Light Chain |
| Applicazioni | Flow Cytometry, Immunohistochemistry (Frozen), Western Blot |
| Classificazione | Monoclonal |
| Clone | ICO-106 |
| Concentrazione | 1 mg/mL |
| Coniugato | Unconjugated |
| Formulazione | PBS with 0.08% sodium azide |
| Specie ospite | Mouse |
| Immunogeno | Human Ig. |
| Metodo di purificazione | Purified |
| Vedi altri risultati |
Titolo del prodotto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?