Learn More
Invitrogen™ Mouse anti-Human Lambda Light Chain Secondary Antibody
Codice prodotto. 11555020
Cambia vista
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
11555020 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11555020 Fornitore Invitrogen™ N. del fornitore SA119254
  • ONLINE EXCLUSIVE PRICE
    209.00€
    181.83€ / 100µg
    Risparmia 27.17€
    13% Sconto
Visualizza tutte le promozioni
 Articolo soggetto a restrizioni
Spedizione stimata: 22-10-2026
Mouse Monoclonal Secondary Antibody
This antibody reacts with lambda light chains (22.5 kDa) of human immunoglobulin.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifica
Antigene Human Lambda Light Chain
Applicazioni ELISA, Western Blot, Immunocytochemistry
Classificazione Monoclonal
Clone Rs4
Concentrazione 1 mg/mL
Coniugato Unconjugated
Formulazione PBS with 15mM sodium azide; pH 7.4
Specie ospite Mouse
Immunogeno Lambda light chains of human immunoglobulin.
Metodo di purificazione Sequential chromatography
Quantità 100 μg
Status giuridico RUO
Primario o secondario Secondary
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Secondary Antibody
Forma Liquid
Isotype IgG1