Learn More
Invitrogen™ Mouse anti-Rat IgG1 (Heavy chain) Secondary Antibody, Biotin
Codice prodotto. 15256059
Cambia vista
Click to view available options
Quantità:
500 μg
Dimensione della confezione:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
15256059 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 15256059 Fornitore Invitrogen™ N. del fornitore MA516815
  • ONLINE EXCLUSIVE PRICE
    762.00€
    662.94€ / 500µg
    Risparmia 99.06€
    13% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 12-11-2026
Mouse Monoclonal Secondary Antibody
Mouse anti rat IgG1, clone MARG1-2 recognizes the heavy chain of the rat immunoglobulin G1 subclass. No cross-reactivity has been found with other rat immunoglobulin classes or subclasses.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifica
Antigene Rat IgG1 (Heavy chain)
Applicazioni ELISA
Classificazione Monoclonal
Clone MARG1-2
Concentrazione 1 mg/mL
Coniugato Biotin
Formulazione PBS with 50% glycerol and 0.1% sodium azide
Specie ospite Mouse
Immunogeno Rat IR27 IgG1 myeloma protein.
Metodo di purificazione Affinity chromatography
Quantità 500 μg
Status giuridico RUO
Primario o secondario Secondary
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Secondary Antibody
Forma Liquid
Isotype IgG1