Learn More
ACROBiosystems Mouse CD3 epsilon&CD3 gamma Heterodimer Protein,Fc,His Tag&Fc,Flag Tag (MALS verified)
Marca: ACROBiosystems CDG-M52W2-1mg
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
T-cell surface glycoprotein CD3 delta & CD3 gamma chain, also known as CD3D & CD3G or CD3D&CD3G respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
Mouse CD3E&CD3G Heterodimer Protein,Fc,His Tag&Fc,Flag Tag is produced by co-expression of CD3E and CD3G, has a calculated MW of 41.5 kDa (CD3E) and 41.6 kDa (CD3G). Subunit CD3E is fused with a human IgG1 Fc tag at the C-terminus, followed by a polyhistiSpecifica
NP_031674.1/NP_033980.1 | |
>95% as determined by SDS-PAGE. | |
Mouse CD3E&CD3G; Heterodimer Protein,Fc,His Tag&Fc;,Flag Tag (CDG-M52W2) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 108 (CD3E) & Gln 23 - Ser 116 (CD3G) (Accession # P22646-1 (CD3E) & P11942-1 (CD3G)). | |
-20°C | |
CD3E & CD3G | |
His Tag & Fc Tag & Flag Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
Mouse |
41.5 kDa/41.6 kDa | |
1 mg | |
Asp 23 - Asp 108 (CD3E) & Gln 23 - Ser 116 (CD3G) | |
CD3 epsilon & CD3 gamma,CD3E & CD3G | |
Unconjugated | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers & CD Antigens |