Learn More
R&D Systems™ Mouse CD8 beta (NP_033988) VersaClone cDNA
Codice prodotto. 18747738
Cambia vista
Click to view available options
Quantità:
10 μg
Dimensione della confezione:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
18747738 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18747738 Fornitore R&D Systems™ N. del fornitore RDC2646
  • ONLINE EXCLUSIVE PRICE
    393.00€
    371.70€ / 10µg
    Risparmia 21.30€
    5% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 30-09-2026
Aggiungi al carrello
Aggiungi al carrello
Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.
  • The Mouse CD8 beta (NP_033988) VersaClone cDNA clone contains the complete ORF for CD8 beta, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The CD8 beta insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
TRUSTED_SUSTAINABILITY
Specifica
Gene mCD8beta
Numero di accesso NP_033988
Concentrazione 10 μg at 0.2 μg/μL
Formulazione cDNA is provided in 10 mM Tris-Cl, pH 8.5
ID gene (immissione) 926
Content And Storage Store the unopened product at -20 to -80°C
Specie Mouse
Quantità 10 μg