Learn More
Mouse CD90/Thy1 Antibody, R&D Systems™
Codice prodotto. 18182781
Cambia vista
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18182781 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18182781 Fornitore R&D Systems N. del fornitore MAB7335
Rat Monoclonal Antibody
CD90/Thy1 Monoclonal specifically detects CD90/Thy1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifica
Antigene CD90/Thy1
Applicazioni Western Blot, Flow Cytometry, CyTOF
Classificazione Monoclonal
Clone 778053
Concentrazione LYOPH
Coniugato Unconjugated
Diluizione Western Blot 0.5 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulazione Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N. accesso geni P01831
Alias gene CD90, CD90 antigen, CDw90, FLJ33325, Thy1, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Simboli geni THY1
Specie ospite Rat
Immunogeno Chinese hamster ovary cell line CHO-derived recombinant mouse CD90/Thy1 Gln20-Lys130 Accession # P01831
Metodo di purificazione Protein A or G purified from hybridoma culture supernatant
Quantità 100 μg
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 7070
Test di specificità Detects mouse CD90/Thy1 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human CD90/Thy1 is observed.
Ricostituzione Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Forma Purified
Isotype IgG2a