Learn More
ACROBiosystems Mouse GUCY2C / Guanylyl cyclase C Protein, Fc Tag
457.25€ - 3357.52€
Specifica
Peso molecolare | 73.5 kDa |
---|---|
Sorgente | Mouse GUCY2C, Fc Tag (GUC-M5254) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Met 433 (Accession # Q3UWA6-1). |
Immunogeno | Val 20 - Met 433 |
Requisiti di stoccaggio | -20°C |
Alias gene | GUCY2C,GUC2C,STAR,STA receptor,hSTAR,GC-C |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
17196351
|
ACROBiosystems
GUC-M5254-100UG |
100 μg |
457.25€
100µg |
Spedizione stimata: 30-04-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
17106361
|
ACROBiosystems
GUC-M5254-1MG |
1 mg |
3357.52€
1mg |
Spedizione stimata: 30-04-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
GUCY2C (Guanylyl Cyclase C), also known as heat-stable enterotoxin receptor, is a type I transmembrane protein of the guanylate cyclase (gc) family that signal by producing cGMP. Guanylate cyclase C (GUCY2C) and its hormones guanylin and uroguanylin have recently emerged as one paracrine axis defending intestinal mucosal integrity against mutational, chemical, and inflammatory injury. GUCY2C murine CAR-T cells recognized and killed human colorectal cancer cells endogenously expressing GUCY2C. Thus, we have identified a human GUCY2C-specific CAR-T cell therapy approach that may be developed for the treatment of GUCY2C-expressing metastatic colorectal cancer.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 73.5 kDa. The protein migrates as 90-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifica
73.5 kDa | |
Val 20 - Met 433 | |
GUCY2C,GUC2C,STAR,STA receptor,hSTAR,GC-C | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
Mouse GUCY2C, Fc Tag (GUC-M5254) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Met 433 (Accession # Q3UWA6-1). | |
-20°C | |
GUCY2C | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |