Learn More
ACROBiosystems Mouse ILDR2 Protein, His Tag
Marca:  ACROBiosystems IL2-M52H5-100ug
Dettagli aggiuntivi : Peso : 0.00010kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17116411
  • dietro preventivo / 100µg
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
Descrizione
Descrizione
ILDR2 belongs to the immunoglobulin superfamily. May be involved in lipid homeostasis and ER stress pathways. ILDR2 as a novel B7-like protein with robust T cell inhibitory activity, expressed in immune cells and in immune-privileged and inflamed tissues. A fusion protein, consisting of ILDR2 extracellular domain with an Fc fragment, that binds to a putative counterpart on activated T cells showed a beneficial effect in the collagen-induced arthritis model and abrogated the production of proinflammatory cytokines and chemokines in autologous synovial-like cocultures of macrophages and cytokine-stimulated T cells. Collectively, these findings point to ILDR2 as a novel negative regulator for T cells, with potential roles in the development of immune-related diseases, including autoimmunity and cancer.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.5 kDa.
Specifica
Specifica
Powder
100 µg
Leu 36 - Glu 201
ILDR2,C1orf32
Unconjugated
HEK293
Mouse
20.5 kDa
Mouse ILDR2, His Tag (IL2-M52H5) is expressed from human 293 cells (HEK293). It contains AA Leu 36 - Glu 201 (Accession # B5TVM2-1).
-20°C
ILDR2
His Tag
Immune Checkpoint Proteins