Learn More
R&D Systems™ Mouse Oct-3/4 (NP_038661.2) VersaClone cDNA
Specifica
Specifica
| Gene | Oct-3/4 |
| Numero di accesso | NP_038661.2 |
| ID gene (immissione) | 5460 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantità | 10 μg |