Learn More
R&D Systems™ Mouse Plexin B3 (NP_062533.2) VersaClone cDNA
Specifica
Specifica
| Gene | Plexin B3 |
| Numero di accesso | NP_062533.2 |
| ID gene (immissione) | 5365 |
| Alias gene | PLEXB3, plexin B3, plexin-B3, PLEXR |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantità | 10 μg |