missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Syndecan-1/CD138 (NP_035649) VersaClone cDNA
Descrizione
- Also known as: Sstn; Synd; CD138; Synd1; syn-1; AA408134; AA409076
- SDC1 is a type I transmembrane heparan sulfate proteoglycan and is a member of the syndecan proteoglycan family.
- The syndecans mediate cell binding, cell signaling, and cytoskeletal organization.
- Syndecan receptors are required for internalization of the HIV-1 tat protein.
- SDC1 functions as an integral membrane protein and participates in cell proliferation, cell migration and cell-matrix interactions via its receptor for extracellular matrix proteins.
- Altered SDC1 expression has been detected in several different tumor types.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifica
Specifica
| Gene | mSdc1 |
| Numero di accesso | NP_035649 |
| Concentrazione | 10 μg at 0.2 μg/μL |
| Formulazione | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Tipo di inserto | 949 bp |
| ID gene (immissione) | 6382 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Specie | Mouse |
| Quantità | 10 μg |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?