Learn More
R&D Systems™ Mouse TGF-beta 3 (NP_033394.2) VersaClone cDNA
Specifica
Specifica
| Gene | TGF-beta 3 |
| Numero di accesso | NP_033394.2 |
| ID gene (immissione) | 7043 |
| Alias gene | ARVD, FLJ16571, TGF-beta3, TGF-beta-3, transforming growth factor beta-3, transforming growth factor, beta 3 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Linea di prodotti | R&D Systems |
| Quantità | 10 μg |