Learn More
MP Biomedicals™ Lysing Matrix Z, 96-Well Rack
Optimized for the FastPrep™-96 Instrument and are included in the FastDNA™-96 Soil Microbe DNA Kit, the FastDNA™-96 Fungal/Bacterial DNA Kit, and the FastDNA™-96 Fecal DNA Kit
Marca:  MP Biomedicals™ 116961001
Dettagli aggiuntivi : Peso : 0.17400kg
 vedi altre versioni di questo prodotto
Codice prodotto. 15292840
  • 195.00€ / Pezzo
Spedizione stimata
15-12-2020
Aggiungi al carrello
Descrizione
Descrizione
  • Each 96 deep well rack comes with 1.2mL cluster-strip tube and cap assemblies. Each tube is pre-filled with 2.0mm diameter Yttria-Stabilized Zirconium Oxide beads.
  • Y2O3 + ZrO2 has high density and has extremely high hardness making this matrix ideal for cracking of very tough cell walls, while it's smooth surface minimizes shearing.
  • Lysing Matrix Z is used primarily for lysis of animal and plant tissue, tougher stems, roots and seeds, and whole insects.
  • Matrix Z is an alternative to the very agressive Matrix A because it's lower shear rate minimizes degradation in applications such as RNA and Protein isolation from very tough tissue.
Specifica
Specifica
cracking of very tough cell walls
FastPrep-96
1.2mL cluster-strip tube prefilled w/2mm dia. Yttria-Stabilized Zirconium Oxide beads
96 x 1.2mL