Learn More
MPP8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA558891
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: GLEHGFEKPL DSAMSAEEDT DVRGRRKKKT PRKAEDTREN RKLENKNAFL EKKTVPKKQR NQDRSKSAAE LEKLMPVSAQ TPKGRRLSGE ERGLWSTDSA EEDKETKRNE SKE Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 58%.
MPP8 Polyclonal antibody specifically detects MPP8 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifica
MPP8 | |
Polyclonal | |
Unconjugated | |
MPHOSPH8 | |
1500035L22Rik; 4930548G07Rik; Ab2-008; AU040635; HSMPP8; M-phase phosphoprotein 8; M-phase phosphoprotein, mpp; M-phase phosphoprotein, mpp8; Mphosph8; MPP8; RGD1305133; TWA3; Two hybrid-associated protein 3 with RanBPM | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54737 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99549 | |
MPHOSPH8 | |
Recombinant protein corresponding to Human MPP8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only