missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ MPure™ Blood DNA Extraction Kit 1200

For extraction and purification of genomic DNA from 400 to 1000μL mammalian whole blood or a suspension of mammalian blood cells

Marca:  MP Biomedicals™ 117022200

Dettagli aggiuntivi : Peso : 1.50000kg

Codice prodotto. 15729210

  • 375.24€ / 1 set
Spedizione stimata: 15-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

  • For extraction of genomic DNA, mitochondrial and viral DNA
  • Compatible with ammalian fresh or frozen whole blood
  • Completely automated purification protocol
  • Runs in as little as 60 minutes
  • Produces nucleic acids that can be used immediately in downstream applications such as PCR, qPCR, Sequencing (NGS), Microarray, RFLP and Southern Blot Analysis

Specifica

Specifica

Store at room temperature
Buffy Coat, Whole Blood Cells
DNA Purification
48 Preps
As little as 60 minutes
Blood DNA Extraction Kit 1200
MPure-12 Automated Nucleic Acid Purification System
Proteinase K solution, 50μL; Lysis Buffer 2, 1500μL; Binding Buffer 1, 1000μL; Magnetic Bead Solution, 1000μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL; Elution Buffer 1, 1000μL; Elution Buffer 2, 1000μL
400 to 1000μL whole blood (WBC count less than 6 x 104 cells/μL); 400 to 1000μL buffy coat (contains no more than 5 x 106 cells)
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali