Learn More
MP Biomedicals™ MPure™ TB DNA Extraction Kit
For extraction of genomic DNA of Mycobacteria spp. (e.g. Mycobacterium tuberculosis) from different specimens
Marca: MP Biomedicals™ 117022800
Dettagli aggiuntivi : Peso : 1.50000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Descrizione
- Variety of biological samples including sputum, bronchoalveolar lavage, pus, blood, cell-free body fluids, urine and respiratory specimens
- Solid or suspended bacterial culture
- Completely automated purification protocol
- Run times as little as 60 minutes
- DNA can be used immediately in downstream applications
Specifica
Store at room temperature | |
Magnetic Bead | |
MPure-12 Automated Nucleic Acid Purification System | |
Proteinase K solution, 40μL; Lysis Buffer 2, 720μL; Binding Buffer 1, 720μL; Magnetic Bead Solution, 800μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL; Elution Buffer 1, 1000μL; Elution Buffer 2, 1000μL; Buffer N1, 400μL | |
As little as 60 minutes |
TB DNA Extraction Kit | |
Mycobacterium Specimens | |
DNA Purification | |
48 Preps |